Diallylamin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

D0069-25ML D0069-500ML
TCIAD0069-25MLEA 222 DKK
TCIAD0069-25ML TCIAD0069-500ML
Diallylamin ≥98.0% (by GC)
Diallylamin
Formel: C₆H₁₁N
Kogepunkt: 112°C
Smeltepunkt: -88°C
Flammepunkt: 14°C
MDL Number: MFCD00008642
CAS-nummer: 124-02-7
EINECS: 204-671-2
UN nummer: 2359
ADR: 3,II
Merck Index: 14,02971
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79b907d0d508a96759d25cabc4381bffb1846cbf&vid=79b907d0d508a96759d25cabc4381bffb1846cbf&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR