Triethylene glycol monomethyl ether ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: mPEG3-Alcohol, 2-(2-(2-Methoxyethoxy)ethoxy)ethanol, Methyl triglycol, m-PEG3-OH

T0709-25ML T0709-500ML
TCIAT0709-25MLEA 140 DKK
TCIAT0709-25ML TCIAT0709-500ML
Triethylene glycol monomethyl ether ≥98.0% (by GC)
Triethylene glycol monomethyl ether
Formel: C₇H₁₆O₄
Molekylvægt: 164.2 g/mol
Kogepunkt: 248°C
Smeltepunkt: –45°C
Flammepunkt: 134°C
MDL Number: MFCD00002875
CAS-nummer: 112-35-6
EINECS: 203-962-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ebd90d0ff453f551c98836978381f1d0045be4b&vid=0ebd90d0ff453f551c98836978381f1d0045be4b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR