Dibenzylphosphonat ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Dibenzyl phosphite

P1016-25G P1016-250G
TCIAP1016-25GEA 411 DKK
TCIAP1016-25G TCIAP1016-250G
Dibenzylphosphonat ≥95.0% (by GC)
Dibenzylphosphonat
Formel: C₁₄H₁₅O₃P
Molekylvægt: 262.25 g/mol
Kogepunkt: 165°C
MDL Number: MFCD00004774
CAS-nummer: 17176-77-1
EINECS: 241-226-1
Merck Index: 14,03014
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41f5b4eeccd764433b30c2cb8ffdb93a7f9367d8&vid=41f5b4eeccd764433b30c2cb8ffdb93a7f9367d8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR