Cysteaminiumchlorid ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-MEA

A0296-25G A0296-500G A0296-100G
TCIAA0296-25GEA 189 DKK
TCIAA0296-25G TCIAA0296-500G TCIAA0296-100G
Cysteaminiumchlorid ≥95.0% (by titrimetric analysis)
Cysteaminiumchlorid
Formel: C₂H₇NS·HCl
Molekylvægt: 113.61 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00012904
CAS-nummer: 156-57-0
EINECS: 205-858-1
Merck Index: 14,02779
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cb100e0a19c16a9e07a4582e7c35400a544d73b&vid=3cb100e0a19c16a9e07a4582e7c35400a544d73b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 95.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR