1,2-Epoxyoctadecan ≥85.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

E0313-25G E0313-400G
TCIAE0313-25GEA 189 DKK
TCIAE0313-25G TCIAE0313-400G
1,2-Epoxyoctadecan ≥85.0% (by GC)
1,2-Epoxyoctadecane
Formel: C₁₈H₃₆O
MDL Number: MFCD00005152
CAS-nummer: 7390-81-0
EINECS: 230-977-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5b90591b45d058251cf05bf69c8c88b94f4850f&vid=e5b90591b45d058251cf05bf69c8c88b94f4850f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR