1,2-Epoxydodecan ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D1984-25ML D1984-100ML
TCIAD1984-25MLEA 132 DKK
TCIAD1984-25ML TCIAD1984-100ML
1,2-Epoxydodecan ≥95.0% (by GC)
1,2-Epoxydodecane
Formel: C₁₂H₂₄O
Kogepunkt: 85°C
Flammepunkt: 105°C
MDL Number: MFCD00005150
CAS-nummer: 2855-19-8
EINECS: 220-667-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce1658bb0fbad6f33c9c29d5c999d4afab7bd3d0&vid=ce1658bb0fbad6f33c9c29d5c999d4afab7bd3d0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR