1,2-Epoxydecan ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
E0315-25ML
E0315-500ML
TCIAE0315-25MLEA
427
DKK
TCIAE0315-25ML
TCIAE0315-500ML
1,2-Epoxydecan ≥97.0% (by GC)
1,2-Epoxydecane
Formel:
C₁₀H₂₀O Kogepunkt: 208°C Flammepunkt: 78°C |
MDL Number:
MFCD00005158 CAS-nummer: 2404-44-6 EINECS: 219-295-4 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...