N,N'-Dimethyl-1,3-propandiamin ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N1,N3-Dimethylpropane-1,3-diamine, N,N′-Dimethyl-1,3-propanediamine

D3811-25G D3811-5G
TCIAD3811-25GEA 1900 DKK
TCIAD3811-25G TCIAD3811-5G
N,N'-Dimethyl-1,3-propandiamin ≥98.0% (by titrimetric analysis)
N,N'-Dimethyl-1,3-propandiamin
Formel: C₅H₁₄N₂
Kogepunkt: 140°C
Flammepunkt: 20°C
MDL Number: MFCD00008292
CAS-nummer: 111-33-1
EINECS: 203-859-1
UN nummer: 2733
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7e1fb221d6d8e22689025542faca49835f2edd1&vid=a7e1fb221d6d8e22689025542faca49835f2edd1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR