N-Methylacetamid ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

M0133-25G M0133-250G
TCIAM0133-25GEA 132 DKK
TCIAM0133-25G TCIAM0133-250G
N-Methylacetamid ≥99.0% (by GC)
N-Methylacetamid
Formel: C₃H₇NO
Kogepunkt: 206°C
MDL Number: MFCD00008683
CAS-nummer: 79-16-3
EINECS: 201-182-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2999c446d8b01f179e04ac98a8b886f46026ec0c&vid=2999c446d8b01f179e04ac98a8b886f46026ec0c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR