Bis(cyclopentadienyl)nikkel ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Nickelocene

D1574-1G D1574-5G
TCIAD1574-1GEA 329 DKK
TCIAD1574-1G TCIAD1574-5G
Bis(cyclopentadienyl)nikkel ≥98.0% (by titrimetric analysis)
Bis(cyclopentadienyl)nikkel
Formel: C₁₀H₁₀Ni
MDL Number: MFCD00001441
CAS-nummer: 1271-28-9
EINECS: 215-039-0
UN nummer: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6cac9b209ebc5874de834f8cc6228952b32467c&vid=d6cac9b209ebc5874de834f8cc6228952b32467c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Dark green to Dark blue to Black powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR