3-Aminoisonicotinsyre ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A2133-1G A2133-5G
TCIAA2133-1GEA 468 DKK
TCIAA2133-1G TCIAA2133-5G
3-Aminoisonicotinsyre ≥98.0% (by HPLC, titration analysis)
3-Aminoisonicotinsyre
Formel: C₆H₆N₂O₂
Smeltepunkt: 303°C
MDL Number: MFCD00137840
CAS-nummer: 7579-20-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fd9c78068206e59042199f5779894e8617da5ff&vid=5fd9c78068206e59042199f5779894e8617da5ff&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR