(Triisopropylsilyl)acetylen ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Ethynyltriisopropylsilane
T1683-25ML
T1683-5ML
TCIAT1683-25MLEA
2110
DKK
TCIAT1683-25ML
TCIAT1683-5ML
(Triisopropylsilyl)acetylen ≥95.0% (by GC)
(Triisopropylsilyl)acetylen
Formel:
C₁₁H₂₂Si Molekylvægt: 182.38 g/mol Kogepunkt: 100°C Flammepunkt: 56°C |
MDL Number:
MFCD00075452 CAS-nummer: 89343-06-6 UN nummer: 1993 ADR: 3 III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...