2-Amino-3,3-dimethylbutan ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A0656-10ML A0656-25ML
TCIAA0656-10MLEA 1470 DKK
TCIAA0656-10ML TCIAA0656-25ML
2-Amino-3,3-dimethylbutan ≥98.0% (by GC)
(±)-3,3-Dimethyl-2-butylamine
Formel: C₆H₁₅N
Kogepunkt: 102°C
Flammepunkt: 1°C
MDL Number: MFCD00008078
CAS-nummer: 3850-30-4
EINECS: 223-353-4
UN nummer: 2733
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f2ea619d7a3287fc6ee0dc77b3e3ed83a76d81c&vid=3f2ea619d7a3287fc6ee0dc77b3e3ed83a76d81c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR