6-Bromisatin ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B2424-25G B2424-5G
TCIAB2424-25GEA 1640 DKK
TCIAB2424-25G TCIAB2424-5G
6-Bromisatin ≥97.0% (by GC, titration analysis)
6-Bromisatin
Formel: C₈H₄BrNO₂
Smeltepunkt: 274°C
MDL Number: MFCD01631138
CAS-nummer: 6326-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23e8e410944e0a64c1ab6d54cc55ead12f84d33b&vid=23e8e410944e0a64c1ab6d54cc55ead12f84d33b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR