N,N-Dimethylpropionamid ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D0793-25ML D0793-500ML D0793-100ML
TCIAD0793-25MLEA 263 DKK
TCIAD0793-25ML TCIAD0793-500ML TCIAD0793-100ML
N,N-Dimethylpropionamid ≥98.0% (by GC)
N,N-Dimethylpropionamid
Formel: C₅H₁₁NO
Molekylvægt: 101.15 g/mol
Kogepunkt: 176°C
Flammepunkt: 62°C
MDL Number: MFCD00009301
CAS-nummer: 758-96-3
EINECS: 212-064-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85054c74239e99b493b7267d3cc5a976e1d3d970&vid=85054c74239e99b493b7267d3cc5a976e1d3d970&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR