Cyclohexen ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
C0491-25ML
C0491-500ML
TCIAC0491-25MLEA
132
DKK
TCIAC0491-25ML
TCIAC0491-500ML
Cyclohexen ≥99.0% (by GC)
Cyclohexene
Formel:
C₆H₁₀ Kogepunkt: 82°C Smeltepunkt: –104 °C Massefylde: 0,81 g/cm³ (20 °C) Flammepunkt: -6°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001539 CAS-nummer: 110-83-8 EINECS: 203-807-8 UN nummer: 2256 ADR: 3,II Merck Index: 14,02727 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...