Bicyclohexyl ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B0902-25ML B0902-500ML B0902-100ML
TCIAB0902-25MLEA 304 DKK
TCIAB0902-25ML TCIAB0902-500ML TCIAB0902-100ML
Bicyclohexyl ≥99.0% (by GC)
Bicyclohexyl
Formel: C₁₂H₂₂
Molekylvægt: 166.31 g/mol
Kogepunkt: 239°C
Flammepunkt: 92°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003815
CAS-nummer: 92-51-3
EINECS: 202-161-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb010a1dfb6c139901043e1827c5565efce0eee7&vid=cb010a1dfb6c139901043e1827c5565efce0eee7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR