3-Methylphthalicanhydrid ≥96.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Methylisobenzofuran-1,3-dione
M1464-1G
M1464-5G
TCIAM1464-1GEA
255
DKK
TCIAM1464-1G
TCIAM1464-5G
3-Methylphthalicanhydrid ≥96.0% (by titrimetric analysis)
3-Methylphthalicanhydrid
Formel:
C₉H₆O₃ |
MDL Number:
MFCD00047316 CAS-nummer: 4792-30-7 EINECS: 225-344-0 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Methanolysis Method) | min. 96.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...