2-Chlorethyl toluen-4-sulfonat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Chloroethyl p-toluenesulphonate

C2380-100G C2380-25G
TCIAC2380-100GEA 1100 DKK
TCIAC2380-100G TCIAC2380-25G
2-Chlorethyl toluen-4-sulfonat ≥98.0% (by GC)
2-Chlorethyl toluen-4-sulfonat
Formel: C₉H₁₁ClO₃S
Kogepunkt: 156°C
MDL Number: MFCD00000970
CAS-nummer: 80-41-1
EINECS: 201-277-2
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c49127d23420b71a5f2f4654951b607b6e4d0b41&vid=c49127d23420b71a5f2f4654951b607b6e4d0b41&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR