Methyl3-furoat ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methyl furan-3-carboxylate
F0468-25G
F0468-5G
TCIAF0468-25GEA
2520
DKK
TCIAF0468-25G
TCIAF0468-5G
Methyl3-furoat ≥96.0% (by GC)
Methyl3-furoat
Formel:
C₆H₆O₃ Kogepunkt: 64°C |
MDL Number:
MFCD06203671 CAS-nummer: 13129-23-2 UN nummer: 3272 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...