Ethyl 3-furoat ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Ethyl furan-3-carboxylate, Ethyl 3-Furoate, Ethyl 3-Furancarboxylate, 3-Furancarboxylic acid ethyl ester, 3-Furoic acid ethyl ester
F0463-25G
F0463-5G
TCIAF0463-25GEA
1420
DKK
TCIAF0463-25G
TCIAF0463-5G
Ethyl 3-furoat ≥98.0% (by GC)
Ethyl 3-furoat
Formel:
C₇H₈O₃ Molekylvægt: 140.14 g/mol Flammepunkt: 59°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00005348 CAS-nummer: 614-98-2 EINECS: 210-403-5 UN nummer: 3272 ADR: 3 III |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...