Cyclododecanol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
C0461-25G
C0461-500G
TCIAC0461-25GEA
132
DKK
TCIAC0461-25G
TCIAC0461-500G
Cyclododecanol ≥98.0% (by GC)
Cyclododecanol
Formel:
C₁₂H₂₄O Molekylvægt: 184.32 g/mol Kogepunkt: 104°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003717 CAS-nummer: 1724-39-6 EINECS: 217-031-2 UN nummer: 3077 ADR: 9 III |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 76.0 to 80.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...