2-Chlorcyclohexanol ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C0656-25G
C0656-5G
TCIAC0656-25GEA
2130
DKK
TCIAC0656-25G
TCIAC0656-5G
2-Chlorcyclohexanol ≥95.0% (by GC)
2-Chlorcyclohexanol
Formel:
C₆H₁₁ClO Kogepunkt: 84°C Massefylde: 1,13 g/cm³ (20 °C) |
MDL Number:
MFCD00003860 CAS-nummer: 1561-86-0 EINECS: 216-339-4 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...