Chlorcyclohexan ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Cyclohexyl chloride
C0153-25ML
C0153-500ML
TCIAC0153-25MLEA
132
DKK
TCIAC0153-25ML
TCIAC0153-500ML
Chlorcyclohexan ≥98.0% (by GC)
Chlorocyclohexane
Formel:
C₆H₁₁Cl Kogepunkt: 143°C Smeltepunkt: -44°C Massefylde: 1 g/cm³ (20 °C) Flammepunkt: 32°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003822 CAS-nummer: 542-18-7 EINECS: 208-806-6 UN nummer: 1993 ADR: 3,III Merck Index: 14,02732 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...