1,4-Cyclohexandion ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C0730-25G
C0730-500G
TCIAC0730-25GEA
304
DKK
TCIAC0730-25G
TCIAC0730-500G
1,4-Cyclohexandion ≥98.0% (by GC)
1,4-Cyclohexanedione
Formel:
C₆H₈O₂ Kogepunkt: 132°C Smeltepunkt: 75…76 °C Massefylde: 1,28 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001606 CAS-nummer: 637-88-7 EINECS: 211-306-0 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...