8-Chlorquinolin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C0284-25G C0284-5G
TCIAC0284-25GEA 3120 DKK
TCIAC0284-25G TCIAC0284-5G
8-Chlorquinolin ≥98.0% (by GC)
8-Chlorquinolin
Formel: C₉H₆ClN
Kogepunkt: 288°C
Smeltepunkt: –20 °C
Massefylde: 1,2834 g/cm³ (14 °C)
MDL Number: MFCD00047618
CAS-nummer: 611-33-6
EINECS: 210-265-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97614a1f8f6b5ebe8dcbcc81793790245fdf611b&vid=e23ab01876d886f1d8111c1d3825ffcae5fd5f0d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR