(+)-3-Carene ≥90.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
C0047-25ML
C0047-500ML
TCIAC0047-25MLEA
140
DKK
TCIAC0047-25ML
TCIAC0047-500ML
(+)-3-Carene ≥90.0% (by GC)
(+)-3-Carene
Formel:
C₁₀H₁₆ Kogepunkt: 170°C Flammepunkt: 46°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00066417 CAS-nummer: 498-15-7 EINECS: 207-856-6 UN nummer: 2319 ADR: 3,III Merck Index: 14,01836 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 90.0 % |
Specific rotation [a]20/D | +13.0 to +17.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...