Cyclohexylbenzen ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Phenylcyclohexane

C0496-25ML C0496-500ML C0496-100ML
TCIAC0496-25MLEA 132 DKK
TCIAC0496-25ML TCIAC0496-500ML TCIAC0496-100ML
Cyclohexylbenzen ≥97.0% (by GC)
Cyclohexylbenzen
Formel: C₁₂H₁₆
Molekylvægt: 160.26 g/mol
Kogepunkt: 236°C
Smeltepunkt: 5
Flammepunkt: 99°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001451
CAS-nummer: 827-52-1
EINECS: 212-572-0
UN nummer: 3082
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29f42f09183cc7c9915055193f442a190e38484f&vid=ec906cd9a6ec9a70fa820c6bf01afcc1b28cb2da&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR