2-Phenylcyclohexanon ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P1311-5G
P1311-1G
TCIAP1311-5GEA
1120
DKK
TCIAP1311-5G
TCIAP1311-1G
2-Phenylcyclohexanon ≥98.0% (by GC)
2-Phenylcyclohexanon
Formel:
C₁₂H₁₄O Molekylvægt: 174.24 g/mol Kogepunkt: 90 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001629 CAS-nummer: 1444-65-1 EINECS: 215-888-7 |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | gt. 57.0 to 61.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...