Cyclohexen-1-ylbenzen ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P1302-25ML P1302-5ML
TCIAP1302-25MLEA 969 DKK
TCIAP1302-25ML TCIAP1302-5ML
Cyclohexen-1-ylbenzen ≥95.0% (by GC)
Cyclohexen-1-ylbenzen
Formel: C₁₂H₁₄
Kogepunkt: 252°C
Smeltepunkt: 0 °C
Massefylde: 1,004 g/cm³ (0 °C)
Flammepunkt: 103°C
MDL Number: MFCD00001542
CAS-nummer: 771-98-2
EINECS: 212-242-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d966d766c789bf4335f8e194ccb34b7733a579c&vid=18b20d2b83fc9bd01b1f1084a9de36a9824ad88b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR