Hexamethylbenzen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H0087-5G H0087-25G
TCIAH0087-5GEA 534 DKK
TCIAH0087-5G TCIAH0087-25G
Hexamethylbenzen ≥99.0% (by GC)
Hexamethylbenzen
Formel: C₁₂H₁₈
Kogepunkt: 266 °C (1013 hPa)
Smeltepunkt: 82…83 °C
Massefylde: 1,063 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008523
CAS-nummer: 87-85-4
EINECS: 201-777-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a77d196fcd33f155de52645b137913e6c8008ca6&vid=a77d196fcd33f155de52645b137913e6c8008ca6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 165.0 to 168.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR