2,4,6-Tris(brommethyl)mesitylen ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1,3,5-Tris(bromomethyl)-2,4,6-trimethylbenzene, 2,4,6-Tris(bromomethyl)mesitylene

T1641-25G T1641-5G
TCIAT1641-25GEA 1430 DKK
TCIAT1641-25G TCIAT1641-5G
2,4,6-Tris(brommethyl)mesitylen ≥98.0% (by titrimetric analysis)
2,4,6-Tris(brommethyl)mesitylen
Formel: C₁₂H₁₅Br₃
Molekylvægt: 398.96 g/mol
MDL Number: MFCD00192554
CAS-nummer: 21988-87-4
EINECS: 244-697-1
UN nummer: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0734c003d664baf18599b660439e8d45979228b&vid=ad5cf8062361d0e884e2a39624e174d5e331c185&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 183.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR