Pentamethylbenzen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,2,3,4,5-Pentamethylbenzene

P0043-25G P0043-5G
TCIAP0043-25GEA 1220 DKK
TCIAP0043-25G TCIAP0043-5G
Pentamethylbenzen ≥99.0% (by GC)
Pentamethylbenzen
Formel: C₁₁H₁₆
Kogepunkt: 232 °C (1013 hPa)
Smeltepunkt: 51…52 °C
Massefylde: 0,917 g/cm³ (20 °C)
MDL Number: MFCD00008522
CAS-nummer: 700-12-9
EINECS: 211-837-8
UN nummer: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd1112be7b1534b29313cb6c34b734a18f0cc247&vid=fd1112be7b1534b29313cb6c34b734a18f0cc247&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 50.0 to 54.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR