1,3-Diethylbenzen ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D0478-25ML D0478-100ML
TCIAD0478-25MLEA 337 DKK
TCIAD0478-25ML TCIAD0478-100ML
1,3-Diethylbenzen ≥97.0% (by GC)
1,3-Diethylbenzen
Formel: C₁₀H₁₄
Molekylvægt: 134.22 g/mol
Kogepunkt: 182°C
Smeltepunkt: –84°C
Massefylde: 0.8798
Flammepunkt: 56°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009260
CAS-nummer: 141-93-5
EINECS: 205-511-4
UN nummer: 2049
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc53b84ae55d34964b4f6b4e9d9eef2462fec804&vid=3da7f8e4498e39d3f5602f73b930bb645bb12cb0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR