3,4-Xylenol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
D0777-25G
D0777-500G
TCIAD0777-25GEA
132
DKK
TCIAD0777-25G
TCIAD0777-500G
3,4-Xylenol ≥98.0% (by GC)
3,4-Xylenol
Formel:
C₈H₁₀O Kogepunkt: 227 °C (1013 hPa) Smeltepunkt: 62…65 °C Massefylde: 1,023 g/cm³ (17 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002304 CAS-nummer: 95-65-8 EINECS: 202-439-5 UN nummer: 2261 ADR: 6.1,II Merck Index: 14,10082 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 63.0 to 68.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...