2,3-Xylenol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2,3-Dimethylphenol
D0773-25G
D0773-500G
TCIAD0773-25GEA
255
DKK
TCIAD0773-25G
TCIAD0773-500G
2,3-Xylenol ≥98.0% (by GC)
2,3-Xylenol
Formel:
C₈H₁₀O Kogepunkt: 217 °C (1013 hPa) Smeltepunkt: 75 °C Massefylde: 0,965 g/cm³ (20 °C) |
MDL Number:
MFCD00002227 CAS-nummer: 526-75-0 EINECS: 208-395-3 UN nummer: 2261 ADR: 6.1,II Merck Index: 14,10082 |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...