3,4-Xylidin ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D0670-25G D0670-500G
TCIAD0670-25GEA 189 DKK
TCIAD0670-25G TCIAD0670-500G
3,4-Xylidin ≥99.0% (by GC, titration analysis)
3,4-Xylidin
Formel: C₈H₁₁N
Kogepunkt: 226°C
Smeltepunkt: 48…51 °C
Massefylde: 1,0755 g/cm³ (18 °C)
Flammepunkt: 98 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007810
CAS-nummer: 95-64-7
EINECS: 202-437-4
UN nummer: 3452
ADR: 6.1,II
Merck Index: 12,10217
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fab5c3a1740157b83d01cbf648631958f50c1b2&vid=65fe6bb9215d219dc40c38a787b9b4823c726480&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 49.0 to 51.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR