3-(Trifluormethyl)benzylchlorid ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-(Trifluoromethyl)benzyl chloride, 3-(Chloromethyl)benzotrifluoride, 1-(Chloromethyl)-3-(trifluoromethyl)benzene

T2290-25G T2290-5G
TCIAT2290-25GEA 1420 DKK
TCIAT2290-25G TCIAT2290-5G
3-(Trifluormethyl)benzylchlorid ≥95.0% (by GC)
3-(Trifluormethyl)benzylchlorid
Formel: C₈H₆ClF₃
Molekylvægt: 194.58 g/mol
Kogepunkt: 70°C
Massefylde: 1.3079
Flammepunkt: 49°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000908
CAS-nummer: 705-29-3
EINECS: 211-884-4
UN nummer: 2920
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d2a137ec1e3b9e04aea9ede27fb128bce3a1fa6&vid=3e9ca41a5fb723e1d0b6ca86cbbb51fec8cf25e3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR