4-(Dimethylamino)benzonitril ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-(Dimethylamino) benzonitrile

D3247-25G D3247-5G
TCIAD3247-25GEA 969 DKK
TCIAD3247-25G TCIAD3247-5G
4-(Dimethylamino)benzonitril ≥98.0% (by GC, titration analysis)
4-(Dimethylamino)benzonitril
Formel: C₉H₁₀N₂
Kogepunkt: 318°C
MDL Number: MFCD00001815
CAS-nummer: 1197-19-9
EINECS: 214-819-8
UN nummer: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9097ba64394de068d7c0c4d10ad0fd88926ba1dc&vid=9097ba64394de068d7c0c4d10ad0fd88926ba1dc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 74.0 to 78.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR