Pentyl nitrite (mixed isomers ) ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Amyl Nitrite, Pentyl Nitrite
A0455-25ML
A0455-500ML
A0455-100ML
TCIAA0455-25MLEA
370
DKK
TCIAA0455-25ML
TCIAA0455-500ML
TCIAA0455-100ML
Pentyl nitrite (mixed isomers ) ≥95.0% (by GC)
Pentyl nitrite (mixed isomers )
Formel:
C₅H₁₁NO₂ Kogepunkt: 104°C Flammepunkt: –18°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00036132 CAS-nummer: 463-04-7 EINECS: 207-332-7 UN nummer: 1113 ADR: 3,II Merck Index: 12,01617 |
|
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...