2-Methoxypropen ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Methoxy-1-propene, Isopropenyl methyl ether, 2-Methoxyprop-1-ene

I0303-25ML I0303-500ML I0303-100ML
TCIAI0303-25MLEA 476 DKK
TCIAI0303-25ML TCIAI0303-500ML TCIAI0303-100ML
2-Methoxypropen ≥95.0% (by GC)
2-Methoxypropene
Formel: C₄H₈O
Kogepunkt: 36°C
Flammepunkt: -25°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00014929
CAS-nummer: 116-11-0
EINECS: 204-125-3
UN nummer: 1993
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98e591658357f5a209ac1402ad281875b246b64a&vid=98e591658357f5a209ac1402ad281875b246b64a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR