4-Butoxyanilin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B1003-25G B1003-5G
TCIAB1003-25GEA 632 DKK
TCIAB1003-25G TCIAB1003-5G
4-Butoxyanilin ≥98.0% (by GC, titration analysis)
4-Butoxyanilin
Formel: C₁₀H₁₅NO
Molekylvægt: 165.24 g/mol
Kogepunkt: 157°C
MDL Number: MFCD00007866
CAS-nummer: 4344-55-2
EINECS: 224-402-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d17aeaab7dee9c0946b0160a656dea4b7b29705a&vid=d17aeaab7dee9c0946b0160a656dea4b7b29705a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR