3,5-Di-tert-butyl-4-hydroxybenzaldehyd ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D1587-25G D1587-500G D1587-100G
TCIAD1587-25GEA 304 DKK
TCIAD1587-25G TCIAD1587-500G TCIAD1587-100G
3,5-Di-tert-butyl-4-hydroxybenzaldehyd ≥98.0% (by HPLC, titration analysis)
3,5-Di-tert-butyl-4-hydroxybenzaldehyd
Formel: C₁₅H₂₂O₂
Kogepunkt: 332 °C (1013 hPa)
Smeltepunkt: 190 °C
Massefylde: 1,16 g/cm³ (20 °C)
MDL Number: MFCD00008826
CAS-nummer: 1620-98-0
EINECS: 216-592-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa709ffe704d0c03847568a820fdbb488f82f8e8&vid=c5c6f7688143e0040ebc0a5f2a2e77b455a70482&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 187.0 to 192.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR