Chlortriisopropylsilan ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T1078-25ML T1078-250ML T1078-5ML
TCIAT1078-25MLEA 682 DKK
TCIAT1078-25ML TCIAT1078-250ML TCIAT1078-5ML
Chlortriisopropylsilan ≥95.0% (by GC)
Chlortriisopropylsilan
Formel: C₉H₂₁ClSi
Kogepunkt: 96°C
Massefylde: 0,9008 g/cm³ (20 °C)
Flammepunkt: 62°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009656
CAS-nummer: 13154-24-0
UN nummer: 2987
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d03420a3a6b000c0a33940d8fec422b04036ec4&vid=5d03420a3a6b000c0a33940d8fec422b04036ec4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR