3,4-Dimethylbenzaldehyd ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D3334-25G D3334-5G
TCIAD3334-25GEA 591 DKK
TCIAD3334-25G TCIAD3334-5G
3,4-Dimethylbenzaldehyd ≥95.0% (by GC)
3,4-Dimethylbenzaldehyde
Formel: C₉H₁₀O
Kogepunkt: 226°C
Massefylde: 1,012 g/cm³ (20 °C)
MDL Number: MFCD00016612
CAS-nummer: 5973-71-7
EINECS: 227-770-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d0cdb33d5e481dcd64a5e999c2000d3a541b310&vid=6ec1150fa4e887d888a3d955b9f63e0d26cd19ba&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR