4-Brom-m-xylen ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,4-Dimethylbromobenzene, 1-Bromo-2,4-dimethylbenzene

B0996-25G B0996-250G
TCIAB0996-25GEA 1460 DKK
TCIAB0996-25G TCIAB0996-250G
4-Brom-m-xylen ≥97.0% (by GC)
4-Brom-m-xylen
Formel: C₈H₉Br
Kogepunkt: 208°C
Smeltepunkt: –1 °C
Massefylde: 1,358 g/cm³ (20 °C)
Flammepunkt: 79°C
MDL Number: MFCD00000071
CAS-nummer: 583-70-0
EINECS: 209-517-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=139b7714b1bf3f5a346a38a8c2fa3cbbf48560eb&vid=d08d24b3f824d2a84798bfd425fe5dc2e332c78e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR