Undecylbenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
U0010-25ML
U0010-5ML
TCIAU0010-25MLEA
936
DKK
TCIAU0010-25ML
TCIAU0010-5ML
Undecylbenzen ≥98.0% (by GC)
Undecylbenzen
Formel:
C₁₇H₂₈ Kogepunkt: 160°C Smeltepunkt: –5 °C Massefylde: 0,855 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00026549 CAS-nummer: 6742-54-7 EINECS: 229-806-2 ADR: 9,III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...