Tetradecylbenzen ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T0866-25G T0866-5G
TCIAT0866-25GEA 1390 DKK
TCIAT0866-25G TCIAT0866-5G
Tetradecylbenzen ≥97.0% (by GC)
Tetradecylbenzen
Formel: C₂₀H₃₄
Kogepunkt: 207°C
Massefylde: 0,8559 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00015080
CAS-nummer: 1459-10-5
EINECS: 215-950-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42bd9895604632d080ad933fa22845551aeb2d31&vid=d2d779f2fa9d2e1a9b9c41721a9ccbbf4eb21347&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR