tert-Pentylbenzen ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A0751-25ML A0751-500ML
TCIAA0751-25MLEA 156 DKK
TCIAA0751-25ML TCIAA0751-500ML
tert-Pentylbenzen ≥97.0% (by GC)
tert-Pentylbenzen
Formel: C₆H₅C(CH₃)₂CH₂CH₃
Kogepunkt: 191°C
Massefylde: 0,8889 g/cm³ (0 °C)
Flammepunkt: 60°C
MDL Number: MFCD00015187
CAS-nummer: 2049-95-8
EINECS: 218-076-0
UN nummer: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da1b5e961f7facd3c049615994115e20388bafbd&vid=38e3681c4328419a3240035ce670bdd88e37a5a5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR