2-Methylanisol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: o-Cresol methyll ether, 2-Methylanisole, 1-Methoxy-2-methylbenzene, 2-Methoxytoluene

M0150-25ML M0150-500ML
TCIAM0150-25MLEA 148 DKK
TCIAM0150-25ML TCIAM0150-500ML
2-Methylanisol ≥98.0% (by GC)
2-Methylanisol
Formel: C₈H₁₀O
Kogepunkt: 171°C
Massefylde: 0,98 g/cm³ (20 °C)
Flammepunkt: 52°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008373
CAS-nummer: 578-58-5
EINECS: 209-426-3
UN nummer: 3271
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58090c178040b7b7a04c744e7aa4708beab4042f&vid=2670e318dd4b268bde0d179787678e134c5043e2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR